Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CELF6
Entrez Gene ID
427479
Gene Name
CUGBP Elav-like family member 6
Gene Aliases
BRUNOL6
Chromosome Location
Chr.10: 2063574 - 2074272 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004943647.3 XP_004943704.3 8 - 9 1044 78
XM_015279063.2 XP_015134549.1 8 - 9 932 78
XM_015279064.2 XP_015134550.1 7 - 8 875 78
XM_015279065.2 XP_015134551.1 7 - 8 873 78
XM_015279066.2 XP_015134552.2 8 - 9 1041 78
XM_025153988.1 XP_025009756.1 7 - 8 787 78
XM_425051.6 XP_425051.5 8 - 9 1044 78
GenBank mRNA EF520347.1 - 8 - 9 921 78

View Full Product Details