Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KBP
Entrez Gene ID
396300
Gene Name
kainate binding protein
Gene Aliases
-
Chromosome Location
Chr.21: 1456666 - 1471680 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205353.2 NP_990684.1 2 - 3 188 69
XM_025142488.1 XP_024998256.1 2 - 3 1009 69
GenBank mRNA X17700.1 - - 180 69

View Full Product Details