Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RASAL3
- Entrez Gene ID
-
101747651
- Gene Name
- RAS protein activator like 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.30: 770477 - 784771 on Build GRCg6a
- Gene Symbol
- WIZ
- Entrez Gene ID
-
426166
- Gene Name
- WIZ zinc finger
- Gene Aliases
-
-
- Chromosome Location
-
Chr.30: 785744 - 835158 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ851711.1 |
- |
17 - 18 |
3060 |
84 |
View Full Product Details