Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CASP18
Entrez Gene ID
693266
Gene Name
initiator caspase
Gene Aliases
-
Chromosome Location
Chr.7: 11436421 - 11444574 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044689.1 NP_001038154.1 4 - 5 846 97
XM_025152263.1 XP_025008031.1 4 - 5 668 97
GenBank mRNA DQ010055.1 - 4 - 5 846 97
EU165363.1 - 3 - 4 558 97
EU165364.1 - 3 - 4 558 97

View Full Product Details