Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRPS2
Entrez Gene ID
418639
Gene Name
phosphoribosyl pyrophosphate synthetase 2
Gene Aliases
-
Chromosome Location
Chr.1: 124459861 - 124485273 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006264.1 NP_001006264.1 1 - 2 171 70
XM_015273663.2 XP_015129149.1 1 - 2 174 70
GenBank mRNA AJ720935.1 - 1 - 2 171 70
CR354205.1 - 1 - 2 154 70

View Full Product Details