Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RER1
Entrez Gene ID
419397
Gene Name
retention in endoplasmic reticulum sorting receptor 1
Gene Aliases
-
Chromosome Location
Chr.21: 1561164 - 1568433 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006300.1 NP_001006300.1 4 - 5 300 87
XM_015296855.2 XP_015152341.1 3 - 4 264 87
GenBank mRNA AJ721109.1 - 4 - 5 300 87
CR387498.1 - 3 - 4 280 87
CR390617.1 - 4 - 5 301 87

View Full Product Details