Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG9
Entrez Gene ID
732654
Gene Name
autophagy protein 9
Gene Aliases
-
Chromosome Location
Chr.19: 42235681 - 42241994 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AM085508.1 - 8 - 9 1515 88

View Full Product Details