Gene Transcripts

Gene Symbol
DHODH
Entrez Gene ID
1723
Gene Name
dihydroorotate dehydrogenase (quinone)
Gene Aliases
DHOdehase, POADS, URA1
Chromosome Location
Chr. 16: 72008744 - 72027659 on Build GRCh38
UniGene ID
Hs.654427
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001361.4 NP_001352.2 7 955
XM_005255827.3 XP_005255884.1 7 1260
XM_005255829.3 XP_005255886.1 6 897
XM_017022990.1 XP_016878479.1 7 1035
GenBank mRNA AK097300.1 - 2 1599
AK292293.1 - 8 1111
BC065245.1 - 7 935 6064723
M94065.1 - 8 937

View Full Product Details