Gene Transcripts

Gene Symbol
GRTP1
Entrez Gene ID
79774
Gene Name
growth hormone regulated TBC protein 1
Gene Aliases
TBC1D6
Chromosome Location
Chr. 13: 113324164 - 113364158 on Build GRCh38
UniGene ID
Hs.745043
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286732.1 NP_001273661.1 4 515
NM_001286733.1 NP_001273662.1 4 515
NM_024719.3 NP_078995.2 4 515
XM_005268333.3 XP_005268390.1 3 352
XM_011537521.2 XP_011535823.1 4 511
XM_011537522.2 XP_011535824.1 4 511
XM_017020755.1 XP_016876244.1 4 473
XM_017020756.1 XP_016876245.1 5 653
GenBank mRNA AB449887.1 - 4 418
AK026127.1 - 2 158
AK300897.1 - 4 465
BC033071.2 - 2 239
BC062461.1 - 3 363
BC080640.1 - 4 472
BC112123.1 - 2 192 8327591
BC112125.1 - 2 204 8327593
BC143682.1 - 2 192
BQ689859.1 - 4 452
CX788865.1 - 3 356

View Full Product Details