Gene Transcripts

Gene Symbol
C12orf45
Entrez Gene ID
121053
Gene Name
chromosome 12 open reading frame 45
Gene Aliases
Chromosome Location
Chr. 12: 104986320 - 104994727 on Build GRCh38
UniGene ID
Hs.295563
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152318.2 NP_689531.2 4 495
GenBank mRNA BC032326.1 - 479
BG829346.1 - 4 496

View Full Product Details