Gene Transcripts
- Gene Symbol
- SEPT12
- Entrez Gene ID
- 124404
- Gene Name
- septin 12
- Gene Aliases
- SPGF10
- Chromosome Location
- Chr. 16: 4777602 - 4791610 on Build GRCh38
- UniGene ID
- Hs.126780
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001154458.2 |
NP_001147930.1
|
6, 7 |
846 |
|
NM_144605.4 |
NP_653206.2
|
7, 8 |
984 |
|
XM_006720846.2 |
XP_006720909.1
|
7, 8 |
1120 |
|
XM_011522379.2 |
XP_011520681.1
|
6, 7 |
633 |
|
XM_017022938.1 |
XP_016878427.1
|
6, 7 |
1098 |
|
GenBank mRNA |
AK058139.1 |
- |
7, 8 |
810 |
|
BC024017.1 |
- |
7, 8 |
784 |
|
BC035619.1 |
- |
7, 8 |
790 |
5172555 |
DQ456996.1 |
- |
6, 7 |
672 |
|
DQ517531.1 |
- |
7, 8 |
861 |
|
EF620906.1 |
- |
7, 8 |
984 |
|
View Full Product Details