Gene Transcripts

Gene Symbol
DCANP1
Entrez Gene ID
140947
Gene Name
dendritic cell associated nuclear protein
Gene Aliases
C5orf20, DCNP1
Chromosome Location
Chr. 5: 135444214 - 135447348 on Build GRCh38
UniGene ID
Hs.152477
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130848.2 NP_570900.1 1 924
GenBank mRNA AB074498.1 - 1 924
BC069608.1 - 1 768 7262210
BC069610.1 - 1 768 7262222
BC074831.2 - 1 710 30915544
BC074832.2 - 1 710 30915326
BC117156.1 - 1 863
BC117158.1 - 1 863

View Full Product Details