Gene Transcripts

Gene Symbol
NKAIN2
Entrez Gene ID
154215
Gene Name
Na+/K+ transporting ATPase interacting 2
Gene Aliases
FAM77B, NKAIP2, TCBA, TCBA1
Chromosome Location
Chr. 6: 123803840 - 124825652 on Build GRCh38
UniGene ID
Hs.656604
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040214.2 NP_001035304.1 2 462
NM_001300737.1 NP_001287666.1 3 526
NM_001300738.1 NP_001287667.1 2 462
NM_001300740.1 NP_001287669.1 2 100
NM_153355.4 NP_699186.2 2 462
XM_011535501.2 XP_011533803.1 2 350
XM_011535502.2 XP_011533804.1 2 117
XM_011535503.2 XP_011533805.1 2 468
XM_017010318.1 XP_016865807.1 2 466
XM_017010319.1 XP_016865808.1 2 350
GenBank mRNA AB070452.1 - 2 271
AK055326.1 - 2 95
AK308927.1 - 2 100
AK309385.1 - 3 157
AK309802.1 - 2 100
AK310763.1 - 2 137
AK315886.1 - 2 101
BC035062.2 - 3 228 5264304
DB174077.1 - 1 137

View Full Product Details