Gene Transcripts

Gene Symbol
CYP4Z1
Entrez Gene ID
199974
Gene Name
cytochrome P450 family 4 subfamily Z member 1
Gene Aliases
CYP4A20
Chromosome Location
Chr. 1: 47067488 - 47118320 on Build GRCh38
UniGene ID
Hs.176588
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178134.2 NP_835235.1 10, 11 1263
GenBank mRNA AK292175.1 - 10, 11 1301
AY262056.1 - 10, 11 1263
AY358631.1 - 10, 11 1376

View Full Product Details