Gene Transcripts
- Gene Symbol
- LOC100996911
- Entrez Gene ID
- 100996911
- Gene Name
- putative ubiquitin-conjugating enzyme E2Q2-like protein
- Gene Aliases
- Chromosome Location
- Chr. Y: 24188880 - 24206168 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC101929235
- Entrez Gene ID
- 101929235
- Gene Name
- putative ubiquitin-conjugating enzyme E2Q2-like protein
- Gene Aliases
- Chromosome Location
- Chr. Y: 25463991 - 25473624 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC105376722
- Entrez Gene ID
- 105376722
- Gene Name
- uncharacterized LOC105376722
- Gene Aliases
- Chromosome Location
- Chr. 15: 84361567 - 84391073 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- UBE2Q2P1
- Entrez Gene ID
- 388165
- Gene Name
- ubiquitin conjugating enzyme E2 Q2 pseudogene 1
- Gene Aliases
- UBE2QP1
- Chromosome Location
- Chr. 15: 84527196 - 84570795 on Build GRCh38
- UniGene ID
- Hs.498348
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003661.2 |
|
1 |
1132 |
|
GenBank mRNA |
AK125266.1 |
- |
1 |
1132 |
|
View Full Product Details