Gene Transcripts
- Gene Symbol
- ADRB1
- Entrez Gene ID
- 153
- Gene Name
- adrenoceptor beta 1
- Gene Aliases
- ADRB1R, B1AR, BETA1AR, RHR
- Chromosome Location
- Chr. 10: 114044047 - 114046908 on Build GRCh38
- UniGene ID
- Hs.99913
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000684.2 |
NP_000675.1
|
1 |
1504 |
|
GenBank mRNA |
J03019.1 |
- |
1 |
1504 |
|
View Full Product Details