Gene Transcripts

Gene Symbol
UBE2C
Entrez Gene ID
11065
Gene Name
ubiquitin conjugating enzyme E2 C
Gene Aliases
UBCH10, dJ447F3.2
Chromosome Location
Chr. 20: 45812576 - 45816957 on Build GRCh38
UniGene ID
Hs.93002
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281741.1 NP_001268670.1 5 478
NM_001281742.1 NP_001268671.1 5 414
NM_007019.3 NP_008950.1 6 619
NM_181799.2 NP_861515.1 6 565
NM_181800.2 NP_861516.1 5 532
NM_181801.3 NP_861517.1 6 656
NR_104036.1 4 327
NR_104037.1 5 422
GenBank mRNA AK311949.1 - 6 586
BC007656.2 - 6 542 3345575
BC016292.1 - 6 548 3910286
BC028161.1 - 5 1158
BC050736.1 - 6 569 6503915
BI858659.1 - 6 656
BM556795.1 - 6 483
BT007300.1 - 6 499
BT020165.1 - 6 499
BT020166.1 - 6 499
BU169315.1 - 6 506
BU171488.1 - 5 438
BU844974.1 - 5 434
CB143441.1 - 4 245
CK904708.1 - 5 444
CN354813.1 - 5 315
CN354815.1 - 5 357
CR456988.1 - 6 499
U73379.1 - 6 539

View Full Product Details