Gene Transcripts

Gene Symbol
PRICKLE2
Entrez Gene ID
166336
Gene Name
prickle planar cell polarity protein 2
Gene Aliases
EPM5
Chromosome Location
Chr. 3: 64093850 - 64445476 on Build GRCh38
UniGene ID
Hs.148105
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198859.3 NP_942559.1 4 959
XM_011533432.2 XP_011531734.1 5 753
XM_011533433.2 XP_011531735.1 5 753
XM_011533434.2 XP_011531736.1 5 578
XM_011533435.2 XP_011531737.1 5 578
XM_011533436.2 XP_011531738.1 4 811
XM_011533437.2 XP_011531739.1 4 9881
XM_011533440.2 XP_011531742.1 5 753
XM_017005798.1 XP_016861287.1 4 683
XM_017005799.1 XP_016861288.1 3 2432
GenBank mRNA AL833539.1 - 4 960
BC119002.1 - 4 940 40082599

View Full Product Details