Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013694.2 | NP_001013716.2 | 3 | 456 | |
XM_011530178.2 | XP_011528480.1 | 3 | 519 | ||
XM_017028799.1 | XP_016884288.1 | 3 | 468 | ||
GenBank mRNA | AK293916.1 | - | 4 | 436 | |
BC066962.1 | - | 4 | 439 | 4820477 | |
DA325985.1 | - | 3 | 451 |