Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286138.1 | NP_001273067.1 | 7 | 1234 | |
NM_006714.4 | NP_006705.1 | 8 | 1448 | ||
XM_005266797.4 | XP_005266854.1 | 8 | 1155 | ||
XM_005266798.2 | XP_005266855.1 | 6 | 1181 | ||
XM_005266799.2 | XP_005266856.1 | 7 | 1323 | ||
XM_005266801.2 | XP_005266858.1 | 6 | 898 | ||
XM_005266802.2 | XP_005266859.1 | 5 | 2836 | ||
XM_017010203.1 | XP_016865692.1 | 5 | 1056 | ||
XM_017010204.1 | XP_016865693.1 | 7 | 1050 | ||
GenBank mRNA | AK000184.1 | - | 8 | 1221 | |
AK301365.1 | - | 7 | 957 | ||
BC018999.2 | - | 8 | 1218 | 3138813 | |
Y08136.1 | - | 3 | 324 |