Gene Transcripts

Gene Symbol
SYNE4
Entrez Gene ID
163183
Gene Name
spectrin repeat containing nuclear envelope family member 4
Gene Aliases
C19orf46, DFNB76, Nesp4
Chromosome Location
Chr. 19: 36003299 - 36008793 on Build GRCh38
UniGene ID
Hs.436743
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039876.2 NP_001034965.1 6 1044
NM_001297735.2 NP_001284664.1 4 705
GenBank mRNA AA991508.1 - 3 328
AK093764.1 - 5 2003
BC052573.1 - 4 695 6568945
CB241656.1 - 2 269

View Full Product Details