The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005340.1 | NP_001005340.1 | 3 - 4 | 529 | 110 |
NM_002510.2 | NP_002501.1 | 3 - 4 | 529 | 110 | |
XM_005249578.2 | XP_005249635.1 | 3 - 4 | 1020 | 110 | |
XM_017011676.1 | XP_016867165.1 | 3 - 4 | 1020 | 110 | |
XM_017011677.1 | XP_016867166.1 | 3 - 4 | 1020 | 110 | |
XM_017011678.1 | XP_016867167.1 | 3 - 4 | 1020 | 110 | |
GenBank mRNA | AF322909.1 | - | 3 - 4 | 427 | 110 |
AJ505015.1 | - | 3 - 4 | 441 | 110 | |
AK292078.1 | - | 3 - 4 | 451 | 110 | |
AK304671.1 | - | 4 - 5 | 510 | 110 | |
AK312279.1 | - | 3 - 4 | 458 | 110 | |
AY359124.1 | - | 3 - 4 | 449 | 110 | |
BC011595.1 | - | 3 - 4 | 417 | 110 | |
BC025297.1 | - | 3 - 4 | 419 | 110 | |
BC032783.1 | - | 3 - 4 | 445 | 110 | |
BT007074.1 | - | 3 - 4 | 368 | 110 | |
X76534.1 | - | 3 - 4 | 459 | 110 |