Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HLX
Entrez Gene ID
3142
Gene Name
H2.0 like homeobox
Gene Aliases
HB24, HLX1
Chromosome Location
Chr.1: 220879401 - 220885059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021958.3 NP_068777.1 2 - 3 1232 69
GenBank mRNA AK127923.1 - 2 - 3 1122 69
AK313298.1 - 2 - 3 1119 69
BC007294.1 - 2 - 3 1217 69
BC033808.1 - 2 - 3 990 69
M60721.1 - 2 - 3 1188 69

View Full Product Details