Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D4
Entrez Gene ID
9882
Gene Name
TBC1 domain family member 4
Gene Aliases
AS160, NIDDM5
Chromosome Location
Chr.13: 75284664 - 75482646 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286658.2 NP_001273587.1 1 - 2 899 82
NM_001286659.2 NP_001273588.1 1 - 2 899 82
NM_014832.4 NP_055647.2 1 - 2 899 82
XM_005266603.1 XP_005266660.1 1 - 2 1377 82
XM_011535331.1 XP_011533633.1 1 - 2 1377 82
GenBank mRNA AB011175.1 - 1 - 2 845 82
AB449885.1 - 1 - 2 498 82
AK307493.1 - 1 - 2 899 82
BC151239.1 - 1 - 2 837 82
FM207106.1 - 1 - 2 498 82
FM207107.1 - 1 - 2 498 82

View Full Product Details