Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HPSE2
Entrez Gene ID
60495
Gene Name
heparanase 2 (inactive)
Gene Aliases
HPA2, HPR2, UFS, UFS1
Chromosome Location
Chr.10: 98457077 - 99235875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166244.1 NP_001159716.1 9 - 10 1363 74
NM_001166245.1 NP_001159717.1 8 - 9 1201 74
NM_001166246.1 NP_001159718.1 10 - 11 1537 74
NM_021828.4 NP_068600.4 10 - 11 1537 74
XM_006717937.2 XP_006718000.1 10 - 11 1597 74
XM_011540030.1 XP_011538332.1 9 - 10 1362 74
XM_011540031.2 XP_011538333.1 9 - 10 1146 74
XM_011540033.2 XP_011538335.1 9 - 10 1170 74
XM_017016495.1 XP_016871984.1 10 - 11 1537 74
XM_017016497.1 XP_016871986.1 9 - 10 1130 74
XM_017016498.1 XP_016871987.1 9 - 10 1431 74
GenBank mRNA AF282885.1 - 8 - 9 1188 74
AF282886.1 - 9 - 10 1350 74
AF282887.1 - 10 - 11 1524 74
AJ299719.1 - 10 - 11 1524 74
AJ299720.1 - 10 - 11 1464 74
AK094037.1 - 9 - 10 1295 74
AK314759.1 - 10 - 11 1537 74
BC112356.1 - 10 - 11 1506 74

View Full Product Details