Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARAP3
Entrez Gene ID
64411
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3
Gene Aliases
CENTD3, DRAG1
Chromosome Location
Chr.5: 141653401 - 141682233 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022481.5 NP_071926.4 32 - 33 4223 85
XM_005268498.1 XP_005268555.1 32 - 33 4127 85
XM_005268499.1 XP_005268556.1 32 - 33 4161 85
XM_006714792.1 XP_006714855.1 31 - 32 3974 85
XM_011537677.2 XP_011535979.1 30 - 31 3642 85
GenBank mRNA AJ310567.1 - 31 - 32 4173 85
AK001579.1 - 15 - 16 2147 85
AK024718.1 - 2 - 3 109 85
AK091385.1 - 31 - 32 3889 85
AK290052.1 - 32 - 33 4216 85
BC009968.2 - 6 - 7 474 85

View Full Product Details