The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006509.3 | NP_006500.2 | 4 - 5 | 635 | 58 |
XM_005259127.3 | XP_005259184.1 | 3 - 4 | 605 | 58 | |
XM_005259128.2 | XP_005259185.1 | 4 - 5 | 635 | 58 | |
GenBank mRNA | AK290594.1 | - | 4 - 5 | 542 | 58 |
AL583128.3 | - | 4 - 5 | 621 | 58 | |
BC028013.1 | - | 4 - 5 | 587 | 58 | |
M83221.1 | - | 4 - 5 | 648 | 58 |