Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFAP2
Entrez Gene ID
4237
Gene Name
microfibrillar associated protein 2
Gene Aliases
MAGP, MAGP-1, MAGP1
Chromosome Location
Chr.1: 16974502 - 16981649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135247.1 NP_001128719.1 1 - 2 74 89
NM_017459.2 NP_059453.1 1 - 2 74 89
GenBank mRNA AK301655.1 - 1 - 2 66 89
AU280150.1 - 1 - 2 94 89

View Full Product Details