Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H12C
Entrez Gene ID
85463
Gene Name
zinc finger CCCH-type containing 12C
Gene Aliases
MCPIP3
Chromosome Location
Chr.11: 110092425 - 110171841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033390.1 NP_203748.1 4 - 5 1199 93
XM_005271715.3 XP_005271772.1 4 - 5 1580 93
XM_011543055.2 XP_011541357.1 4 - 5 1312 93
XM_017018477.1 XP_016873966.1 4 - 5 5750 93
GenBank mRNA AB051513.1 - 2 - 3 372 93
AB096241.1 - 3 - 4 1936 93
AK295437.1 - 4 - 5 1200 93
AK299100.1 - 4 - 5 1232 93

View Full Product Details