Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C8orf46
Entrez Gene ID
254778
Gene Name
chromosome 8 open reading frame 46
Gene Aliases
-
Chromosome Location
Chr.8: 66493256 - 66518524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152765.3 NP_689978.2 4 - 5 740 76
GenBank mRNA AK055047.1 - 1 - 2 74 76
AK096336.1 - 5 - 6 528 76
AK308800.1 - 5 - 6 567 76
AK315486.1 - 4 - 5 483 76
BC028400.2 - 4 - 5 464 76

View Full Product Details