The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139281.2 | NP_644810.1 | 23 - 23 | 4956 | 98 |
GenBank mRNA | AK055664.1 | - | 1 - 1 | 549 | 98 |
AL832494.1 | - | 21 - 21 | 4666 | 98 | |
CR749211.1 | - | 18 - 18 | 4155 | 98 |