The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197131.1 | NP_001184060.1 | 5 - 6 | 449 | 67 |
NM_002481.3 | NP_002472.2 | 22 - 23 | 2961 | 67 | |
NM_032103.2 | NP_115286.1 | 10 - 11 | 1316 | 67 | |
NM_032104.2 | NP_115287.1 | 10 - 11 | 1316 | 67 | |
XM_017001342.1 | XP_016856831.1 | 23 - 24 | 3144 | 67 | |
XM_017001343.1 | XP_016856832.1 | 23 - 24 | 3141 | 67 | |
XM_017001344.1 | XP_016856833.1 | 23 - 24 | 3144 | 67 | |
XM_017001345.1 | XP_016856834.1 | 23 - 24 | 3141 | 67 | |
XM_017001346.1 | XP_016856835.1 | 22 - 23 | 2961 | 67 | |
XM_017001347.1 | XP_016856836.1 | 22 - 23 | 2958 | 67 | |
XM_017001348.1 | XP_016856837.1 | 22 - 23 | 2958 | 67 | |
XM_017001350.1 | XP_016856839.1 | 23 - 24 | 3018 | 67 | |
XM_017001352.1 | XP_016856841.1 | 13 - 14 | 1411 | 67 | |
XM_017001353.1 | XP_016856842.1 | 13 - 14 | 1401 | 67 | |
XM_017001354.1 | XP_016856843.1 | 10 - 11 | 2795 | 67 | |
XM_017001355.1 | XP_016856844.1 | 6 - 7 | 2145 | 67 | |
GenBank mRNA | AB003062.1 | - | 22 - 23 | 2974 | 67 |
AB050641.1 | - | 10 - 11 | 1316 | 67 | |
AB050642.1 | - | 10 - 11 | 1316 | 67 | |
AB588820.1 | - | 5 - 6 | 449 | 67 | |
AF324888.1 | - | 22 - 23 | 2955 | 67 | |
BC144699.1 | - | 22 - 23 | 2953 | 67 |