The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322468.1 | NP_001309397.1 | - | 21157 | 55 |
NM_004532.5 | NP_004523.3 | 16 - 17 | 2431 | 55 | |
NM_018406.6 | NP_060876.5 | - | 15139 | 55 | |
NM_138297.4 | NP_612154.2 | 15 - 16 | 2278 | 55 | |
GenBank mRNA | AJ010901.1 | - | 16 - 17 | 2215 | 55 |
AJ242541.1 | - | 17 - 18 | 2440 | 55 | |
AJ242542.1 | - | 15 - 16 | 2140 | 55 | |
AJ242543.1 | - | 16 - 17 | 2139 | 55 | |
AJ242544.1 | - | 16 - 17 | 2111 | 55 | |
AJ242545.1 | - | 15 - 16 | 2050 | 55 | |
AJ242546.1 | - | 14 - 15 | 1785 | 55 | |
AJ242547.1 | - | 16 - 17 | 2215 | 55 | |
AJ242548.1 | - | 12 - 13 | 1537 | 55 | |
AJ242549.1 | - | 16 - 17 | 2315 | 55 | |
AJ242550.1 | - | 15 - 16 | 2162 | 55 | |
AJ276359.1 | - | 18 - 19 | 5325 | 55 | |
AJ277412.1 | - | 17 - 18 | 5169 | 55 | |
AJ277505.1 | - | 16 - 17 | 5066 | 55 | |
AJ400633.1 | - | 18 - 19 | 5339 | 55 | |
AJ400849.1 | - | 18 - 19 | 5249 | 55 | |
AJ400850.1 | - | 18 - 19 | 5249 | 55 | |
AJ400852.1 | - | 17 - 18 | 5093 | 55 | |
AJ400853.1 | - | 17 - 18 | 5065 | 55 | |
AJ400854.1 | - | 17 - 18 | 5170 | 55 | |
AJ400855.1 | - | 17 - 18 | 5141 | 55 | |
AJ400856.1 | - | 18 - 19 | 5296 | 55 | |
AJ400857.1 | - | 18 - 19 | 5325 | 55 | |
AJ400858.1 | - | 18 - 19 | 5325 | 55 |