Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR36
Entrez Gene ID
134430
Gene Name
WD repeat domain 36
Gene Aliases
GLC1G, TA-WDRP, TAWDRP, UTP21
Chromosome Location
Chr.5: 111092172 - 111130502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139281.2 NP_644810.1 10 - 11 1374 120
GenBank mRNA AF385437.1 - 10 - 11 1374 120
AK074202.1 - 4 - 5 424 120
AK095312.1 - 10 - 11 1232 120
AL832494.1 - 9 - 10 1176 120
BC133025.1 - 10 - 11 1334 120
BC136517.1 - 10 - 11 1334 120
CR749211.1 - 5 - 6 574 120

View Full Product Details