Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR36
Entrez Gene ID
134430
Gene Name
WD repeat domain 36
Gene Aliases
GLC1G, TA-WDRP, TAWDRP, UTP21
Chromosome Location
Chr.5: 111092172 - 111130502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139281.2 NP_644810.1 17 - 18 2194 71
GenBank mRNA AF385437.1 - 17 - 18 2194 71
AK074202.1 - 11 - 12 1244 71
AK095312.1 - 17 - 18 2052 71
AL832494.1 - 16 - 17 1996 71
BC133025.1 - 17 - 18 2154 71
BC136517.1 - 17 - 18 2154 71

View Full Product Details