Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSBPL6
Entrez Gene ID
114880
Gene Name
oxysterol binding protein like 6
Gene Aliases
ORP6
Chromosome Location
Chr.2: 178194479 - 178399433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201480.1 NP_001188409.1 14 - 15 1909 59
NM_001201481.1 NP_001188410.1 12 - 13 1741 59
NM_032523.3 NP_115912.1 13 - 14 1834 59
NM_145739.2 NP_665682.1 12 - 13 1422 59
XM_011510539.1 XP_011508841.1 13 - 14 1812 59
XM_017003265.1 XP_016858754.1 15 - 16 1981 59
XM_017003266.1 XP_016858755.1 15 - 16 1981 59
XM_017003267.1 XP_016858756.1 13 - 14 1676 59
XM_017003270.1 XP_016858759.1 13 - 14 1895 59
XM_017003273.1 XP_016858762.1 11 - 12 1644 59
GenBank mRNA AB208898.1 - 14 - 15 1909 59
AF323728.1 - 13 - 14 1668 59
AF392448.1 - 13 - 14 1668 59
AK027600.1 - 6 - 7 709 59
AK093902.1 - 12 - 13 1424 59

View Full Product Details