The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172477.1 | NP_001165948.1 | 2 - 3 | 430 | 66 |
NM_015713.4 | NP_056528.2 | 2 - 3 | 449 | 66 | |
GenBank mRNA | AB036063.1 | - | 2 - 3 | 449 | 66 |
AB166671.1 | - | 2 - 3 | 205 | 66 | |
AK001965.1 | - | 2 - 3 | 244 | 66 | |
AK304354.1 | - | 2 - 3 | 374 | 66 | |
AL137348.1 | - | 2 - 3 | 251 | 66 | |
BC108261.1 | - | 2 - 3 | 261 | 66 | |
BC117496.1 | - | 2 - 3 | 286 | 66 | |
BC130628.1 | - | 2 - 3 | 286 | 66 | |
CR627376.1 | - | 2 - 3 | 374 | 66 | |
DC308409.1 | - | 2 - 3 | 430 | 66 |