The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128165.1 | NP_001121637.1 | 2 - 3 | 502 | 116 |
NM_153214.2 | NP_694946.2 | 2 - 3 | 502 | 116 | |
XM_006712260.3 | XP_006712323.1 | 2 - 3 | 502 | 116 | |
XM_011510585.2 | XP_011508887.1 | 2 - 3 | 502 | 116 | |
XM_011510587.2 | XP_011508889.1 | 3 - 4 | 479 | 116 | |
XM_017003317.1 | XP_016858806.1 | 2 - 3 | 502 | 116 | |
XM_017003318.1 | XP_016858807.1 | 2 - 3 | 502 | 116 | |
XM_017003319.1 | XP_016858808.1 | 2 - 3 | 502 | 116 | |
GenBank mRNA | AK094759.1 | - | 2 - 3 | 502 | 116 |
BC035784.1 | - | 2 - 3 | 455 | 116 | |
BC126986.1 | - | 2 - 3 | 304 | 116 | |
BC126987.1 | - | 2 - 3 | 304 | 116 | |
CR933697.1 | - | 2 - 3 | 2116 | 116 | |
DA201691.1 | - | 2 - 3 | 502 | 116 |