The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014335.2 | NP_055150.1 | 1 - 1 | 372 | 58 |
GenBank mRNA | AF092135.1 | - | 1 - 1 | 350 | 58 |
AF109873.1 | - | 1 - 1 | 350 | 58 | |
AF349444.1 | - | 1 - 1 | 342 | 58 | |
AK098383.1 | - | 2 - 2 | 261 | 58 | |
AK315365.1 | - | 1 - 1 | 356 | 58 | |
AL109701.1 | - | 1 - 1 | 312 | 58 | |
AL357456.1 | - | 1 - 1 | 300 | 58 | |
BC114944.1 | - | 1 - 1 | 364 | 58 | |
BC114946.1 | - | 1 - 1 | 364 | 58 | |
BI550673.1 | - | 1 - 1 | 377 | 58 |