The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020318.2 | NP_064714.2 | 4 - 5 | 3299 | 81 |
NM_021936.2 | NP_068755.2 | 4 - 5 | 3299 | 81 | |
XM_005245422.3 | XP_005245479.1 | 4 - 5 | 3346 | 81 | |
XM_011509857.1 | XP_011508159.1 | 5 - 6 | 3216 | 81 | |
XM_011509858.2 | XP_011508160.1 | 4 - 5 | 3559 | 81 | |
XM_017002023.1 | XP_016857512.1 | 5 - 6 | 3397 | 81 | |
XM_017002024.1 | XP_016857513.1 | 5 - 6 | 3444 | 81 | |
XM_017002025.1 | XP_016857514.1 | 4 - 5 | 3550 | 81 | |
GenBank mRNA | AF311940.1 | - | 3 - 4 | 2135 | 81 |
AF342989.1 | - | 4 - 5 | 3299 | 81 | |
AF342990.1 | - | 4 - 5 | 3299 | 81 | |
AJ278348.1 | - | 3 - 4 | 2098 | 81 | |
BC117193.1 | - | 3 - 4 | 2907 | 81 | |
BC117195.1 | - | 3 - 4 | 2908 | 81 | |
BC152552.1 | - | 4 - 5 | 3287 | 81 |