The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308245.1 | NP_001295174.1 | 2 - 3 | 342 | 62 |
NM_152547.4 | NP_689760.2 | 2 - 3 | 342 | 62 | |
XM_006714823.3 | XP_006714886.1 | 2 - 3 | 338 | 62 | |
XM_006714824.3 | XP_006714887.1 | 2 - 3 | 338 | 62 | |
XM_011534444.2 | XP_011532746.1 | 2 - 3 | 338 | 62 | |
XM_017009113.1 | XP_016864602.1 | 2 - 3 | 338 | 62 | |
XM_017009115.1 | XP_016864604.1 | 2 - 3 | 338 | 62 | |
GenBank mRNA | AK057097.1 | - | 2 - 3 | 330 | 62 |
AY358358.1 | - | 2 - 3 | 328 | 62 | |
BC062459.1 | - | 2 - 3 | 201 | 62 | |
DC379841.1 | - | 2 - 3 | 342 | 62 |