The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253693.1 | NP_001240622.1 | 6 - 7 | 806 | 139 |
NM_018298.10 | NP_060768.8 | 7 - 8 | 974 | 139 | |
XM_005271003.1 | XP_005271060.1 | 8 - 9 | 1371 | 139 | |
XM_006710750.1 | XP_006710813.1 | 8 - 9 | 1190 | 139 | |
XM_011541739.2 | XP_011540041.2 | 7 - 8 | 898 | 139 | |
XM_011541740.2 | XP_011540042.1 | 6 - 7 | 732 | 139 | |
XM_011541741.2 | XP_011540043.1 | 7 - 8 | 905 | 139 | |
XM_011541742.1 | XP_011540044.1 | 8 - 9 | 1108 | 139 | |
GenBank mRNA | AF475085.1 | - | 7 - 8 | 880 | 139 |
AK001868.1 | - | 6 - 7 | 712 | 139 | |
AK292206.1 | - | 7 - 8 | 906 | 139 | |
AK314354.1 | - | 7 - 8 | 934 | 139 | |
BC060765.1 | - | 8 - 9 | 1204 | 139 | |
BX647497.1 | - | 9 - 10 | 1127 | 139 |