The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152636.2 | NP_689849.2 | 6 - 7 | 1230 | 66 |
GenBank mRNA | AK091298.1 | - | 5 - 6 | 882 | 66 |