The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144937.1 | NP_001138409.1 | 9 - 10 | 1880 | 96 |
GenBank mRNA | AK093157.1 | - | 5 - 6 | 1206 | 96 |
BC057762.1 | - | 5 - 6 | 1253 | 96 | |
BC130421.1 | - | 5 - 6 | 1206 | 96 |