The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144877.2 | NP_001138349.1 | 10 - 11 | 1086 | 76 |
NM_173690.4 | NP_775961.2 | 11 - 12 | 1155 | 76 | |
GenBank mRNA | AK093983.1 | - | 10 - 11 | 1045 | 76 |
AK098024.1 | - | 11 - 12 | 1092 | 76 | |
BC104030.2 | - | 11 - 12 | 1083 | 76 | |
BC104031.1 | - | 10 - 11 | 1014 | 76 | |
BC104032.1 | - | 10 - 11 | 1014 | 76 |