The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000158.3 | NP_000149.3 | 1 - 2 | 424 | 142 |
GenBank mRNA | AB209731.1 | - | 1 - 2 | 314 | 142 |
AK125918.1 | - | 1 - 2 | 787 | 142 | |
AK312245.1 | - | 1 - 2 | 273 | 142 | |
BC012098.1 | - | 1 - 2 | 353 | 142 | |
L07956.1 | - | 1 - 2 | 231 | 142 |