Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GBE1
Entrez Gene ID
2632
Gene Name
glucan (1,4-alpha-), branching enzyme 1
Gene Aliases
APBD, GBE, GSD4
Chromosome Location
Chr.3: 81489699 - 81761799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000158.3 NP_000149.3 9 - 10 1517 64
GenBank mRNA AB209731.1 - 9 - 10 1407 64
AK125918.1 - 9 - 10 1880 64
AK297939.1 - 7 - 8 950 64
AK300620.1 - 9 - 10 1164 64
AK312245.1 - 9 - 10 1366 64
BC012098.1 - 9 - 10 1446 64
L07956.1 - 9 - 10 1324 64

View Full Product Details