The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182519.2 | NP_872325.2 | 14 - 15 | 1757 | 79 |
GenBank mRNA | AF549190.1 | - | 13 - 14 | 1625 | 79 |