The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182519.2 | NP_872325.2 | 5 - 6 | 940 | 141 |
GenBank mRNA | AF549190.1 | - | 4 - 5 | 808 | 141 |