Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNF16
Entrez Gene ID
7564
Gene Name
zinc finger protein 16
Gene Aliases
HZF1, KOX9
Chromosome Location
Chr.8: 144930358 - 144950888 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029976.2 NP_001025147.2 4 - 4 2327 58
NM_006958.2 NP_008889.2 3 - 3 2241 58
XM_005272341.2 XP_005272398.1 3 - 3 2218 58
XM_005272342.2 XP_005272399.1 3 - 3 2326 58
XM_011517298.1 XP_011515600.1 3 - 3 2343 58
GenBank mRNA AF244088.1 - 2 - 2 2152 58
AK127625.1 - 3 - 3 2241 58
AK226016.1 - 3 - 3 2211 58
AL834451.1 - 2 - 2 2158 58
BC010996.2 - 3 - 3 2175 58
DQ117529.1 - 4 - 4 2327 58

View Full Product Details